Learn More
Medchemexpress LLC HY-112829 50mg , Denifanstat CAS:1399177-37-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11282950MG
Encompass_Preferred
HY-112829 50mg Denifanstat CAS: 1399177-37-7 Denifanstat (TVB-2640) is an orally active and potent Fatty Acid Synthase (FASN) inhibitor with an IC50 of 0.052 μM and an EC50 of 0.072 μM. Denifanstat has the potential for fatty liver disease and cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9951