Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP7858EC10UG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: His-SUMO. Host: E. coli. Species Reactivity: Human. Protein Expression: A DNA sequence encoding the Homo sapiens (Human) DENN domain-containing protein 1A, was expressed in the hosts and tags indicated. AA Sequence: See company website. Buffer: Tris-based buffer, 50% glycerol. Purity: Greater than 90% as determined by SDS-PAGE. Endotoxin: Not determined. Gene Symbol: DENND1A. Gene ID: 57706. Uniprot: Q8TEH3.
Catalog No. 50-188-7357