Learn More
Aviva Systems Biology Corp DENND1B Antibody - N-terminal region (ARP52770_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52770_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DENND1B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DENND1B. Alternate names: FAM31B, MGC27044, C1ORF18, C1orf218, RP11-53I24.2ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60826