Learn More
Medchemexpress LLC DENR Human His-SUM 50ug
SDP
Catalog No. 5000183356
Encompass_Preferred
Recombinant human DENR protein supplied as an N-terminal 6xHis and SUMO fusion, expressed in Escherichia coli and purified for use in biochemical and molecular biology research. The product is suitable for protein-protein interaction studies, functional assays, and assay development, with purity confirmed by reducing SDS-PAGE and guidance for reconstitution provided by the manufacturer.
  • N-terminal 6xHis and SUMO tag.
  • Expressed in Escherichia coli.
  • Derived from human sequence (accession O43583).
  • Purity greater than 90% by reducing SDS-PAGE.
  • Available in 5, 10, 50, and 100 μg sizes.
  • Reconstitute at 100 μg/mL or higher in ddH2O.
Catalog No. 50-001-83356 Supplier Medchemexpress LLC Supplier No. HYP7217250UG
Recombinant human DENR protein supplied as an N-terminal 6xHis and SUMO fusion, expressed in Escherichia coli and purified for use in biochemical and molecular biology research. The product is suitable for protein-protein interaction studies, functional assays, and assay development, with purity confirmed by reducing SDS-PAGE and guidance for reconstitution provided by the manufacturer.
  • N-terminal 6xHis and SUMO tag.
  • Expressed in Escherichia coli.
  • Derived from human sequence (accession O43583).
  • Purity greater than 90% by reducing SDS-PAGE.
  • Available in 5, 10, 50, and 100 μg sizes.
  • Reconstitute at 100 μg/mL or higher in ddH2O.