Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Dense Storage Options
GREENER_CHOICE

Catalog No. 12566298 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Cryobank Tubes
Cryobox
Cryobox Divider
Undivided CryoBox
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
12-566-298 Cryobank Tubes
12-565-004 Cryobox Divider
15-350-55 Undivided CryoBox
15-350-501 Cryobox
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12-566-298 Supplier Thermo Scientific™ Supplier No. 374115
  • $2,580.00
    $1,401.65 / Case of 845

    Save $1,178.35

    46% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Double your cryobank capacity and cut storage costs by 50%. Our dense storage options fit standard cryogenic storage boxes that work with the freezer racks you currently own.

Double your cryobank storage capacity and cut storage costs by 50%. The Thermo Scientific™ Dense Storage Options use standard CryoBoxes™ with 13 x 13 tube separators and fit 169 1mL tubes in a single CryoBox™. These standard-sized boxes are compatible with existing standard-sized lab freezer racks.

  • Choose a standard-sized cardboard or polycarbonate box
  • Insert the 13x13 box divider
  • Load the box with 169, 1mL Cryobank storage tubes
  • Slide the box into your existing standard-sized CryoBox racks
  • Suitable for mechanical freezer or vapor phase liquid nitrogen storage
TRUSTED_SUSTAINABILITY

Specifications

Description 1.0mL Nunc Cryobank Tubes, Blank, with Caps, Sterile, Loose Packed
Coding None
Vial Type Cryobank
Color White, Clear (Closure)
Packaging 169 tubes/bag
For Use With (Equipment) Cryobank
Quantity 845/Cs.
Sterility Sterile
Type Cryobank Tubes
Capacity (Metric) 1.0 mL
Holds 196 1 mL Nunc Cryobank Tubes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.