Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Density Ball
GREENER_CHOICESCI_ED

Catalog No. S139422 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
6+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139422 6+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139422 Supplier Supertek™ Supplier No. PH10088A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Demonstrate the effect of temperature on density of a liquid with this Supertek™ Density Ball.

  • Consists of a 3 in. stainless steel hollow sphere.
  • Weighted so it will float when placed in cool water (15°C) and sink in warm water (30°C).
  • Instructions include 3 experiments.

Specifications

Grade 6+
Format Kit
Languages English
Subject Physcial Science - Density
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.