Learn More
Scientific Industries Density Kit for Torbal AGCN AGZN Scales
Catalog No. 502529293
Encompass_Preferred
The Torbal Density Determination Kit is designed to allow Torbal Analytical Balances to be used to determine the density of solids and fluids using Archimedes Principle in a way that eliminates the need for below the pan weighing. Torbal also makes available an Excel spreadsheet which makes the calculations required for density determinations. (Analytical Balance not included and sold seperately)
Catalog No. 50-252-9293 Supplier Scientific Industries Supplier No. 301067
The Torbal Density Determination Kit is designed to allow Torbal Analytical Balances to be used to determine the density of solids and fluids using Archimedes Principle in a way that eliminates the need for below the pan weighing. Torbal also makes available an Excel spreadsheet which makes the calculations required for density determinations. (Analytical Balance not included and sold seperately)