Learn More
METTLER TOLEDO™ Density Kit, XPR/XSR-Analytical
Catalog No. 13940069 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Application):
Measuring density
Catalog No. For Use With (Application)
13-940-069 Measuring density
1 options
Catalog No. 13-940-069 Supplier METTLER TOLEDO™ Supplier No. 30460852
Add to Cart
Edge
Add to Cart
Kit used to determine density of liquid or solids. (Additional accessories needed if using for liquids)
  • Density Kits are easily installed on Excellence Balances for fast and precise density determination of solid, liquid, porous and pasty substances
  • Fully integrated workflows for five different methods: The built-in density application guides you step-by-step through the procedure
  • The application software collects the density values, calculates the results and allows statistical evaluation of the measurement series
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Measuring density
Type Density Kit