Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Density: Lava Lamp
GREENER_CHOICESCI_ED

Catalog No. s42858 Shop All Aldon Products
Change view
Click to view available options
Class Size:
5 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S42858 5 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

WARNING: This product can expose you to chemicals including Acetyl Salicylic Acid/ Aspirin, which is known to the State of California to cause birth defects and reproductive harm.

CHOKING HAZARD - Small parts. Not for children under 3 yrs.

Catalog No. S42858 Supplier Innovating Science™ Supplier No. IS1008
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Acetyl Salicylic Acid/ Aspirin, which is known to the State of California to cause birth defects and reproductive harm.

CHOKING HAZARD - Small parts. Not for children under 3 yrs.

Make your own lava lamp while learning. Innovating Science™ Density: Lava Lamp is ideal for teaching the concepts of denisty.

  • The concept of density can be difficult to explain to elementary students
  • Help them visualize and understand density and introduce the idea of polarity as well with this demonstration for the whole classroom

Specifications

Certifications/Compliance Product SDSs
Color White
Grade Level Elementary
Grade 2 to 5
Age 10 to 13
For Use With (Application) Science Education
Includes 5 Plastic bottles, 1 L Vegetable Oil, 25 mL Food Coloring, 10 Alka Seltzer tablets
Quantity 1
Dimensions (L x W x H) 9 x 9 x 9 in.
Class Size 5 Lab Groups
Product Type General Chemistry
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.