Learn More
Roche Diagnostics DENSITY REF ST BEADS BATCH A
Supplier:  Roche Diagnostics 06422659001
Encompass
The Density Reference Standard Beads (DRSB) are designed to mimic cell behavior in flow dynamics. Due to their size and optical properties, they will be detected as dead cells by the Cedex Software.
Catalog No. 50-589-382