Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSC Go Science Crazy Density Rod Set
SCI_ED

Catalog No. s28111 Shop All GSC Products
Encompass
Change view
Click to view available options
Includes:
5 rods
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S28111 5 rods
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S28111 Supplier GSC Go Science Crazy Supplier No. DNRDST05
Only null left
Add to Cart
Add to Cart

Density rod sets are ideal for studying the relationship between mass and density.

  • Five rods have equal mass but different lengths
  • Rods are made of PVC, brass, polyethylene, aluminum and nylon
  • Diameter: 12mm

Specifications

Product Type Density rod set
Includes 5 rods
Material PVC, Brass, Polyethylene, Aluminum, Nylon
For Use With (Application) Study the relationship between mass and density
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.