Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Density, Set of 10 Cubes
GREENER_CHOICESCI_ED

Catalog No. S139419 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
6+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139419 6+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139419 Supplier Supertek™ Supplier No. PH10040
Add to Cart
Add to Cart

This Supertek™ Density Set of 10 Cubes consists of one inch density cubes in a storage box.

  • Consists of 10 - 1 in. density cubes of different material.
  • Acrylic
  • Aluminum
  • Brass
  • Copper
  • Nylon
  • Maple
  • Poplar
  • Pine
  • Merandi
  • Steel
  • Cubes stationed in a sturdy foam insert.

Specifications

Grade 6+
Format Set
Languages English
Subject Physcial Science - Density
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.