Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Density, Set of 18 Blocks
SCI_ED

Catalog No. S139421 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
6+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139421 6+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy

Prop 65 Warning

Catalog No. S139421 Supplier Supertek™ Supplier No. PH10051A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Prop 65 Warning

This Supertek™ Density Set of 18 Blocks is excellent for experimenting with density and volume.

  • Includes 18 density blocks of various sizes and materials.
  • Each size has metals of aluminum, brass, copper, lead, steel, and zinc.
  • 6- 10 mm cubes
  • 6- 20 mm cubes
  • 6- 30 x 20 x 10 mm blocks
  • Blocks are nested in sturdy foam within a clear box.

Specifications

Grade 6+
Format Set
Languages English
Subject Physcial Science - Density

California Proposition 65 Warning:
This product contains chemicals known to the State of California to cause cancer, and birth defects or other reproductive harm.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.