Learn More
Sigma Aldrich Fine Chemicals Biosciences Density Standard 20 C UKAS
Supplier:  Sigma Aldrich Fine Chemicals Biosciences DEN201360ML
Encompass
This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.
Catalog No. 50-290-1670