Learn More
Sigma Aldrich Fine Chemicals Biosciences DENSITY STANDARD (20 C)
Supplier:  Sigma Aldrich Fine Chemicals Biosciences DEN201260ML
Encompass
Density Standard (20 °C)(UKAS ISO/IEC17025 and ISO Guide 34 certified), density: 1.1917 g/mL at 20 °C
Catalog No. NC1495127