Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Density Standard (749 kg/m3 HD Fitzgerald Ltd. Quality), MilliporeSigma™ Supelco™

Catalog No. 111018469 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8469 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8469 Supplier MilliporeSigma Supelco Supplier No. 7608110ML
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages, Personal Care and Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 401°F
CAS 112-40-3
Vapor Pressure 1 mmHg (47.8°C)
Vapor Density 5.96 (vs. air)
Density 0.75 g/mL (at 25°C)
Flash Point 83°C - Closed Cup
Linear Formula CH3(CH2)10 CH3
MDL Number MFCD00008969
Melting Point -9.6°C
Boiling Point 215°C to 217°C
Synonym Dodecane
Type Density Standard
Quantity 10 mL
Formula Weight 170.33
Grade H&D; Fitzgerald Ltd. Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.