Learn More
Med Vet International Dental Curette Barnhart 5/6
Catalog No. 502232499
Encompass_Preferred
Curettes are used to remove subgingival calculus, for root planing and for curettage. They have one blade with two cutting edges. Both cutting edges may be used on the front and back of the tooth.
This item is not returnable. View return policy
Catalog No. 50-223-2499 Supplier Med Vet International Supplier No. DB#5/6
Curettes are used to remove subgingival calculus, for root planing and for curettage. They have one blade with two cutting edges. Both cutting edges may be used on the front and back of the tooth.