Learn More
Stoelting™ Dental Cement
SDP
Catalog No. 10000786 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Pink, opaque
Catalog No. Model
10-000-786 Pink, opaque
1 options
Catalog No. 10-000-786 Supplier Stoelting™ Supplier No. 51458
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Dentel Cement is used to affix stainless steel to skull or bone, for lab repairs, or for casting of hard acrylic parts.
  • Particularly useful for securing bone anchor screws in skull holes or cementing cannulae in place
  • Available only to the 48 contiguous US via ground shipment
Specifications
For Use With (Application) Neuroscience
Size 1 lb.
Model Pink, opaque