Learn More
Med Vet International Dental Elevator with Notched "V" Tip, J0899S, Set of 4
Catalog No. 5000583989
Encompass_Preferred
Modified with a "V" cut into the end tip of this instrument, this helps the tip stay on the tooth surface and guides into the socket, preventing it from slipping and damaging the gingiva. The curved tip is designed to follow the root curves of teeth, (i.e., canine and incisor teeth). The elevator tip can be on either the outside or inside of the curved end.
Catalog No. 50-005-83989 Supplier Med Vet International Supplier No. JOR899S
Modified with a "V" cut into the end tip of this instrument, this helps the tip stay on the tooth surface and guides into the socket, preventing it from slipping and damaging the gingiva. The curved tip is designed to follow the root curves of teeth, (i.e., canine and incisor teeth). The elevator tip can be on either the outside or inside of the curved end.