Learn More
Med Vet International Miltex Dental Witzel Root Splinter Extracting Forceps, 6.5in., Upper Roots
Catalog No. 502832099
Encompass_Preferred
The Miltex DEF120 Witzel splinter extracting forceps are made of OR grade German stainless steel and measure 6.5" in length. They have non-locking plier handles and jaws designed for upper roots.
Catalog No. 50-283-2099 Supplier Med Vet International Supplier No. MILDEF120
The Miltex DEF120 Witzel splinter extracting forceps are made of OR grade German stainless steel and measure 6.5" in length. They have non-locking plier handles and jaws designed for upper roots.