Learn More
Med Vet International Dental Hand Instrument Tray 3.625" x 7.75", J0943
Catalog No. 5000583932
Encompass_Preferred
Organizes and stores up to six hand instruments. Autoclavable up to 270 degrees farenheit.
Catalog No. 50-005-83932 Supplier Med Vet International Supplier No. JOR943
Organizes and stores up to six hand instruments. Autoclavable up to 270 degrees farenheit.