Learn More
Med Vet International Dental Insert - Universal Tip (JOR841 Dental Unit only), J0841D3
Catalog No. 5000584934
Encompass_Preferred
This insert is for the J0841 Dental Unit only. They will not work with J0841q or J0841x. Ultrasonic Dental Insert. Universal tip.NOTE: These inserts are NOT interchangeable with any other dental units.
Catalog No. 50-005-84934 Supplier Med Vet International Supplier No. JOR841D3
This insert is for the J0841 Dental Unit only. They will not work with J0841q or J0841x. Ultrasonic Dental Insert. Universal tip.NOTE: These inserts are NOT interchangeable with any other dental units.