Learn More
Med Vet International Miltex Dental Jacquette Scaler, 2/3, Double-Ended, Octagonal Handle
Catalog No. 502830594
Encompass_Preferred
The Miltex 70-100 Jacquette 2/3 dental scaler is made of OR grade German stainless steel. It has an octagonal handle and is double-ended. This scaler can be used to remove supragingival calculus and deposits.
Catalog No. 50-283-0594 Supplier Med Vet International Supplier No. MIL70100
The Miltex 70-100 Jacquette 2/3 dental scaler is made of OR grade German stainless steel. It has an octagonal handle and is double-ended. This scaler can be used to remove supragingival calculus and deposits.