Learn More
Med Vet International Miltex Dental Scaler, H6/7, Double-Ended, Octagonal Handle
Catalog No. 502830622
Encompass_Preferred
The Miltex 70-126 H6/7 dental scaler is made of OR grade German stainless steel. It has an octagonal handle and is double-ended. This scaler can be used to remove supragingival calculus and deposits.
Catalog No. 50-283-0622 Supplier Med Vet International Supplier No. MIL70126
The Miltex 70-126 H6/7 dental scaler is made of OR grade German stainless steel. It has an octagonal handle and is double-ended. This scaler can be used to remove supragingival calculus and deposits.