Learn More
Med Vet International Miltex Dental Scaler #3, Single-Ended, Octagonal Handle
Catalog No. 502830625
Encompass_Preferred
The Miltex 70-108 #3 dental scaler is made of OR grade German stainless steel. It has an octagonal handle and is single-ended. This scaler can be used to remove supragingival calculus and deposits.
Catalog No. 50-283-0625 Supplier Med Vet International Supplier No. MIL70108
The Miltex 70-108 #3 dental scaler is made of OR grade German stainless steel. It has an octagonal handle and is single-ended. This scaler can be used to remove supragingival calculus and deposits.