Learn More
Med Vet International Miltex Dental Whiteside Scaler, 1-0, Single-Ended, Octagonal Handle
Catalog No. 502830627
Encompass_Preferred
The Miltex 70-116 Whiteside 1-0 scaler is made of OR grade German stainless steel. It has an octagonal handle is single-ended. This scaler can be used to remove supragingival calculus and deposits.
Catalog No. 50-283-0627 Supplier Med Vet International Supplier No. MIL70116
The Miltex 70-116 Whiteside 1-0 scaler is made of OR grade German stainless steel. It has an octagonal handle is single-ended. This scaler can be used to remove supragingival calculus and deposits.