Learn More
Med Vet International Miltex Dental Scaler, U15/30, Double-Ended, Octagonal Handle
Catalog No. 502830628
Encompass_Preferred
The Miltex 70-120 U15/30 dental scaler is made of OR grade German stainless steel. It has an octagonal handle and is double-ended. This scaler can be used to remove supragingival calculus and deposits.
Catalog No. 50-283-0628 Supplier Med Vet International Supplier No. MIL70120
 Restricted Item
Add to Cart
Add to Cart
The Miltex 70-120 U15/30 dental scaler is made of OR grade German stainless steel. It has an octagonal handle and is double-ended. This scaler can be used to remove supragingival calculus and deposits.