Learn More
Polysciences, Inc. Dental wax; Low melting; Sheet size: 6 x 3 x 1/16 ft. (150 x 75 x 1.5mm) 1LB
SDP
Catalog No. NC9600249
Encompass
Dental wax; Low melting; Sheet size: 6 x 3 x 1/16 ft. (150 x 75 x 1.5mm); Hazards: HarmLess-use normal precautions; Handling: Exercise normal care; Storage: Storage: RT
Catalog No. NC9600249 Supplier Polysciences, Inc. Supplier No. 004031
Dental wax; Low melting; Sheet size: 6 x 3 x 1/16 ft. (150 x 75 x 1.5mm); Hazards: HarmLess-use normal precautions; Handling: Exercise normal care; Storage: Storage: RT