Learn More
Medchemexpress LLC (-)-Denudatin B | 87402-88-8 | 1 MG
SDP
Catalog No. 5000420378
Encompass_Preferred
Denudatin B is an antiplatelet agent that relaxes vascular smooth muscle. It functions by inhibiting calcium ion influx through voltage-gated and receptor-operated calcium channels, and also exhibits a nonspecific antiplatelet action. This compound is provided with a purity of 97.88%.
  • Acts as an antiplatelet agent
  • Relaxes vascular smooth muscle
  • Inhibits calcium ion influx through voltage-gated and receptor-operated calcium channels
  • Exhibits nonspecific antiplatelet action
  • Stored at 4°C, sealed, away from moisture and light
  • In solvent, stored at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-20378 Supplier Medchemexpress LLC Supplier No. HYN37291MG
Denudatin B is an antiplatelet agent that relaxes vascular smooth muscle. It functions by inhibiting calcium ion influx through voltage-gated and receptor-operated calcium channels, and also exhibits a nonspecific antiplatelet action. This compound is provided with a purity of 97.88%.
  • Acts as an antiplatelet agent
  • Relaxes vascular smooth muscle
  • Inhibits calcium ion influx through voltage-gated and receptor-operated calcium channels
  • Exhibits nonspecific antiplatelet action
  • Stored at 4°C, sealed, away from moisture and light
  • In solvent, stored at -80°C for 6 months or -20°C for 1 month