Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Denudatin B | 87402-88-8 | 5 MG
SDP

Catalog No. 5000420212
Encompass_Preferred

Denudatin B is an antiplatelet agent that relaxes vascular smooth muscle by inhibiting Ca2+ influx through voltage-gated and receptor-operated Ca2+ channels. It exhibits a nonspecific antiplatelet action.

  • Antiplatelet agent
  • Relaxes vascular smooth muscle
  • Inhibits Ca2+ influx through voltage-gated and receptor-operated Ca2+ channels
  • Nonspecific antiplatelet action
  • Molecular formula: C21H24O5
  • Molecular weight: 356.41
  • Target: Calcium channel
  • Pathway: Membrane transporter/ion channel; neuronal signaling
  • Soluble in DMSO (50 mg/mL)

Catalog No. 50-004-20212 Supplier Medchemexpress LLC Supplier No. HYN37295MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Denudatin B is an antiplatelet agent that relaxes vascular smooth muscle by inhibiting Ca2+ influx through voltage-gated and receptor-operated Ca2+ channels. It exhibits a nonspecific antiplatelet action.

  • Antiplatelet agent
  • Relaxes vascular smooth muscle
  • Inhibits Ca2+ influx through voltage-gated and receptor-operated Ca2+ channels
  • Nonspecific antiplatelet action
  • Molecular formula: C21H24O5
  • Molecular weight: 356.41
  • Target: Calcium channel
  • Pathway: Membrane transporter/ion channel; neuronal signaling
  • Soluble in DMSO (50 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.