Learn More
Sino Biological Dengue virus (DENV) (type 2, strain New Guinea C/PUO-218 hybrid) E / Envelope Protein Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 40471T6220
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Dengue virus (DENV) (type 2, strain New Guinea C/PUO-218 hybrid) E / Envelope Protein ( Catalog#40471-V08B; AAC59274.1; Phe247-Gly675). Dengue virus (DENV) (type 2, strain New Guinea C/PUO-218 hybrid) E / Envelope Protein specific IgG was purified by Dengue virus (DENV) (type 2, strain New Guinea C/PUO-218 hybrid) E / Envelope Protein affinity chromatography.
Catalog No. 50-241-2452