Learn More
Sino Biological DENV DENV-E PAb 20UL 40471-T46-20
Supplier:  Sino Biological 40471T4620
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Dengue virus (DENV) (type 2, strain New Guinea C/PUO-218 hybrid) E / Envelope Protein ( Catalog#40471-V08Y1; AAC59274.1; Met578-Gln680). Dengue virus (DENV) (type 2, strain New Guinea C/PUO-218 hybrid) E / Envelope Protein specific IgG was purified by Dengue virus (DENV) (type 2, strain New Guinea C/PUO-218 hybrid) E / Envelope Protein affinity chromatography.
Catalog No. 50-241-2172