Learn More
Sino Biological DENV DENV-NS1 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 40263MM0220
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Dengue virus (DENV) (type 2, strain New Guinea C) NS1 ( Catalog#40263-V07H; AAC59275.1; Asp1-Ala352). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-3194