Learn More
Sino Biological DENV Envelope / DENV-E Protein (His Tag)
Supplier:  Sino Biological 40531V08H100
Encompass_Preferred
A DNA sequence encoding the Dengue virus (DENV)(type 1, strain US/Hawaii/1944) E (ACF49259.1) (Val580-Lys680) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3707