Learn More
Sino Biological Dengue virus (DENV) (type 4, strain Philippines/H241/1956) E / Envelope Protein (aa 579-679, His Tag)
Supplier:  Sino Biological 40533V08H100
Encompass_Preferred
A DNA sequence encoding the envelope protein of Dengue virus (DENV)(type 4, strain Philippines/H241/1956) Envelope Protein (AAX48017.1) (Thr579-Lys679) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3706