Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

InBios International Inc. DENV Detect™ IgM Capture ELISA
SDP

Catalog No. 22022645
Change view
Click to view available options
Target Species:
Human
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Target Species
22-022-645 Human
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-022-645 Supplier InBios International Inc. Supplier No. DDMS1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The DENV Detect™ IgM Capture ELISA is for the qualitative detection of IgM antibodies to DENV recombinant antigens in serum for the presumptive clinical laboratory diagnosis of Dengue virus infection.

  • US FDA-cleared for in vitro diagnostic use and CE Marked
  • Performance thoroughly evaluated with clinically confirmed cases of dengue 1-4 serotypes
  • Relative Sensitivity: >79%, Relative Specificity: >90% (in patients without other flavivirus infections)
  • Strippable plates

Specifications

Immunoassay Kit Format 96-well Strip Plate
Product Type Diagnostic Test
Sample Type Serum
Target Species Human
Storage Requirements 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.