Learn More
Sartorius Balance Automobile Cigarette Lighter Adapter
Catalog No. 01913354
Change view
Click to view available options
Adapter Autolighter 6ft For Xl
Catalog No.
01-913-354 Adapter Autolighter 6ft For Xl
1 options
Catalog No. 01-913-354 Supplier Sartorius Supplier No. 900027.1
Includes
a 6 ft. long (1.8m) cable.
Automobile Cigarette Lighter Adapter. Operate any DI (except Model DI-12K), TL, TR, TC, XL, XE, or XS balance with a standard automobile cigarette lighter. For 12V negative ground automobile systems.