Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius™ Series 200 Benchtop Research-Grade Multiparameter Meters

Catalog No. 02226161
Change view
Click to view available options
Voltage:
115 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
02-226-161 115 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-226-161 Supplier Sartorius™ Supplier No. 9368.1

missing translation for 'validMainframeMsg'

Includes

Meter Only option includes power supply, electrode support arm and operating manual. Meter Kit option consists of the meter, power supply, electrode support arm, operating manual, and a high-performance glass-body pH/ATC electrode.

Research-grade meters for multiparameter measurements

Easy-to-operate models for multiple measurements
Model 220 displays both pH and conductivity measurements at one time
Model 225 is ideal for ion-selective analyses, pH and mV measurements
Model 250 is the multimeter for electrochemical analyses of concentration, conductivity, TDS, salinity, pH and mV
(For the most accurate pH measurement (±0.002), choose Model 215, described under Electrochemistry/pH Meters. )
Single-button, menu-driven operation

  • MODE selects pH, mV, ion, or conductivity; press STANDARDIZE to standardize your measurements
  • CAL DATA displays complete calibration data, including buffers/standards, time/date, slope, cell constant, and calibration line graphs
  • CHANNEL allows you to see one to three parameters at one time
  • Use SETUP to control meter operations
  • DATALOG sets up datalogging and viewing of stored measurements
  • CLEAR cancels an input or returns to previous screen
  • ENTER/PRINT enters selections or prints current results. “Soft keys” provide quick access to the most frequently used features, including context-sensitive help
  • Enter standards and parameters with the numeric keypad
All models have RS-232-C bidirectional input/output
  • Output results with sample alphanumeric description or ID number, result, units, time, date, temperature, channel and stability indication
  • Print at programmed time intervals or by serial port command.
  • Calibration points are time- and date-stamped, and electrode/cell calibration information can be recalled and printed for GLP documentation
  • Meters automatically prompt when recalibration is needed.
  • Internal datalog stores up to 620 results for viewing or downloading
  • Multiline 53/4 in. backlit display shows time, date, current buffers, temperature, electrode slope, and current value; prompts for input data

Model Selection
Model 220

  • For simultaneous pH and conductivity measurements, even with electrodes in the same solution
  • Switches from conductivity to NaCl salinity, practical salinity, resistivity, or Total Dissolved Solids
  • Automatically selects the correct range for each sample from five subranges or autorange units, or uses fixed units in μS/cm or kilohm•cm
  • Features automatic conductivity calibration, allows up to five points for calibration, and uses advanced four-band cell conductivity circuitry for the most accurate conductivity measurements available\
  • Temperature correction can be programmed either off or on, with coefficients from 0 to 4.0% per °C
Model 225
  • Displays direct ISE readouts in ppt, ppm, ppb, %, mg/mL, μg/L, moles/L, and generic ion units
  • Select one of 14 preprogrammed ions and enter up to seven standards, using fixed- or floating-point notation
  • Meter guides you through eight different known-addition or subtraction methods for indirect ion measurement
  • Dual channels provide research-grade pH and mV measurements simultaneously
  • Built-in buffer and temperature tables provide automatic compensation with the ATC probe (included in the glass-body pH/ATC probe)
Model 250
  • Measures pH, mV, temperature, conductivity, salinity, resistivity, total dissolved solids and ion concentration
  • Automatically selects the correct range for the sample and calibrates cell constant
  • Uses up to five standards
  • Exceeds USP 23 conductivity protocol requirements
  • Displays direct ion readout in ppm, ppb, %, mg/L, μg/L, mole/L and generic ion units, with units in use displayed
  • Choose from 14 different ions with units and get eight incremental modes for indirect concentration measurements

Order Info

Meter dimensions: 8L x 12 1/4W x 4 in.H (20 x 31 x 10cm). Weight: 3 1/4 lb. (2.1kg). For 115V 50/60Hz.

Specifications

Includes Meter, power supply, electrode support arm and operating manual
Product Type Multiparameter Meter
Voltage 115 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.