Learn More
Thomas Scientific Mini Vortexer, Blue Cup Head, 100 to 240V (US Plug)
SDP
Catalog No. 501461263
Encompass
Powerful vortexing for tubes up to 50ml Insta-Touch pressure activated operation "Mini Sized", less than 4 in. wide Available in five color configurations Powerful is the best word to describe this mini vortexer. With a 4mm orbit and fixed speed of 2800rpm, the labForce Mini Vortexer instantly vortexes even the largest samples, including nearly full 50 ml tubes. Despite its powerful motor, the Mini Vortexer has a footprint of less than 4x4 inches, allowing it to fit on even the most crowded bench. Insta-Touch operation means that the motor is automatically activated with slight pressure on the center cup head. Operation is smooth, quiet and efficient. When pressure is removed, operation immediately ceases. The Mini Vortexer's unique head design prevents liquids from entering the housing, prolonging motor life. The unit is available with five different cup head colors.
Catalog No. 50-146-1263 Supplier Thomas Scientific Supplier No. 1154J88
Powerful vortexing for tubes up to 50ml Insta-Touch pressure activated operation "Mini Sized", less than 4 in. wide Available in five color configurations Powerful is the best word to describe this mini vortexer. With a 4mm orbit and fixed speed of 2800rpm, the labForce Mini Vortexer instantly vortexes even the largest samples, including nearly full 50 ml tubes. Despite its powerful motor, the Mini Vortexer has a footprint of less than 4x4 inches, allowing it to fit on even the most crowded bench. Insta-Touch operation means that the motor is automatically activated with slight pressure on the center cup head. Operation is smooth, quiet and efficient. When pressure is removed, operation immediately ceases. The Mini Vortexer's unique head design prevents liquids from entering the housing, prolonging motor life. The unit is available with five different cup head colors.