Learn More
Medchemexpress LLC Deoxy-bigchap | MFCD20924625 | 99.9% | 862.06 | C42H75N3O15 | 100 MG
SDP
Catalog No. 5000281344
Encompass_Preferred
Deoxy-bigchap is a non-ionic detergent used as a biochemical reagent for solubilization and gentle extraction of membrane proteins and other biomolecules in life-science research. Supplied with analytical documentation, it is intended for research use only and supports reproducible experimental workflows.
  • Non-ionic detergent for membrane solubilization.
  • Improved solubility compared with related detergents.
  • Suitable for gentle extraction of membrane proteins.
  • Provided with analytical documents (SDS, COA, HNMR, RP-HPLC, MS).
  • High purity to support reproducible results.
Catalog No. 50-002-81344 Supplier Medchemexpress LLC Supplier No. HYW250173100MG
Deoxy-bigchap is a non-ionic detergent used as a biochemical reagent for solubilization and gentle extraction of membrane proteins and other biomolecules in life-science research. Supplied with analytical documentation, it is intended for research use only and supports reproducible experimental workflows.
  • Non-ionic detergent for membrane solubilization.
  • Improved solubility compared with related detergents.
  • Suitable for gentle extraction of membrane proteins.
  • Provided with analytical documents (SDS, COA, HNMR, RP-HPLC, MS).
  • High purity to support reproducible results.