Learn More
Cayman Chemical Deoxy Donepezil (hydrochloride), Purity: ≥98%, CAS: 1034439-57-0, 10 mg
SDP
Catalog No. 502989929
Encompass_Preferred
A potential impurity found in bulk preparations of donepezil; is produced as a by-product during donepezil synthesis
Catalog No. 50-298-9929 Supplier Cayman Chemical Supplier No. 3004510 MG
A potential impurity found in bulk preparations of donepezil; is produced as a by-product during donepezil synthesis