Learn More
Medchemexpress LLC Deoxy-thalidomide-5-iodine | 2291364-01-5 | MFCD32853043 | 98.0% | 370.14 g/mol | C13H11IN2O3 | 250 MG
SDP
Catalog No. 5000281760
Encompass_Preferred
Deoxy-thalidomide-5-iodine is a thalidomide analog used as a cereblon-binding ligand in targeted protein degradation and PROTAC research. It is provided as a characterized small-molecule building block suitable for conjugation in bifunctional degrader synthesis and related chemical biology applications.
  • Used as cereblon-binding ligand for PROTAC development.
  • High reported purity: 98.0% (NMR).
  • Molecular weight 370.14 g/mol and formula C13H11IN2O3.
  • Solid; store at 4°C and protect from light.
  • Available in multiple package sizes, including 250 mg.
Catalog No. 50-002-81760 Supplier Medchemexpress LLC Supplier No. HYW441271250MG
Deoxy-thalidomide-5-iodine is a thalidomide analog used as a cereblon-binding ligand in targeted protein degradation and PROTAC research. It is provided as a characterized small-molecule building block suitable for conjugation in bifunctional degrader synthesis and related chemical biology applications.
  • Used as cereblon-binding ligand for PROTAC development.
  • High reported purity: 98.0% (NMR).
  • Molecular weight 370.14 g/mol and formula C13H11IN2O3.
  • Solid; store at 4°C and protect from light.
  • Available in multiple package sizes, including 250 mg.