Learn More
Apexbio Technology LLC Deoxyarbutin 53936-56-4 50mg
SDP
Catalog No. 502240345
Encompass_Preferred
Deoxyarbutin (CAS 53936-56-4) is a small molecule that acts as a reversible inhibitor of tyrosinase a key enzyme involved in melanin biosynthesis By targeting tyrosinase Deoxyarbutin effectively suppresses enzymatic activity exhibiting an IC50 value of 50 nM in inhibition assays This compound is widely utilized in biomedical research to study melanogenesis explore regulatory mechanisms of skin pigmentation and evaluate potential therapeutic approaches for pigmentary disorders
This item is not returnable. View return policy
Catalog No. 50-224-0345 Supplier Apexbio Technology LLC Supplier No. B192550
Deoxyarbutin (CAS 53936-56-4) is a small molecule that acts as a reversible inhibitor of tyrosinase a key enzyme involved in melanin biosynthesis By targeting tyrosinase Deoxyarbutin effectively suppresses enzymatic activity exhibiting an IC50 value of 50 nM in inhibition assays This compound is widely utilized in biomedical research to study melanogenesis explore regulatory mechanisms of skin pigmentation and evaluate potential therapeutic approaches for pigmentary disorders