Learn More
Medchemexpress LLC Deoxyarbutin | 53936-56-4 | 99.9% | 100 G
SDP
Supplier:  Medchemexpress LLC HYB1461100G
Encompass_Preferred
Deoxyarbutin is an orally active tyrosinase inhibitor. It has been shown to promote apoptosis in melanoma cells, enhance the vitality of mouse acinar cells, and exhibits skin whitening and anti-tumor activities. It is intended for research use only.
  • Orally active tyrosinase inhibitor.
  • Promotes apoptosis of melanoma cells.
  • Enhances the vitality of mouse acinar cells.
  • Possesses skin whitening activity.
  • Demonstrates anti-tumor activity.
  • Low cytotoxicity to normal cell lines in vitro.
  • Exhibits a dose-dependent skin whitening effect in hairless pigmentary guinea pigs.
  • Inhibits tumor growth and lung metastasis in a melanoma mouse model.
  • Can alleviate severe acute pancreatitis in mice.
Catalog No. 50-004-12700