Learn More
Medchemexpress LLC Deoxyarbutin | 53936-56-4 | 99.92% | 10 G
SDP
Catalog No. 5000412636
Encompass_Preferred
Deoxyarbutin is an orally active tyrosinase inhibitor that promotes apoptosis of melanoma cells, enhances the vitality of mouse acinar cells, and exhibits skin whitening and anti-tumor activity.
  • Promotes melanoma cell apoptosis through the p38-mediated mitochondrial related apoptosis pathway.
  • Restores mitochondrial function and enhances cell viability in mouse pancreatic acinar cells.
  • Inhibits the proliferation of tumor cells (B16F10 and LL/2 cells) with lower cytotoxicity to normal cell lines.
  • Exhibits a dose-dependent skin whitening effect.
  • Demonstrates anti-tumor activity and anti-tumor metastasis in melanoma mice.
  • Alleviates severe acute pancreatitis in mice via the HtraA2/PGC-1a pathway.
Catalog No. 50-004-12636 Supplier Medchemexpress LLC Supplier No. HYB146110G
Add to Cart
Add to Cart
Deoxyarbutin is an orally active tyrosinase inhibitor that promotes apoptosis of melanoma cells, enhances the vitality of mouse acinar cells, and exhibits skin whitening and anti-tumor activity.
  • Promotes melanoma cell apoptosis through the p38-mediated mitochondrial related apoptosis pathway.
  • Restores mitochondrial function and enhances cell viability in mouse pancreatic acinar cells.
  • Inhibits the proliferation of tumor cells (B16F10 and LL/2 cells) with lower cytotoxicity to normal cell lines.
  • Exhibits a dose-dependent skin whitening effect.
  • Demonstrates anti-tumor activity and anti-tumor metastasis in melanoma mice.
  • Alleviates severe acute pancreatitis in mice via the HtraA2/PGC-1a pathway.