Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Deoxybrevianamide E | 34610-68-9 | MFCD14635384 | >99.0% | 351.44 g/mol | C21H25N3O2 | 1MG
SDP

Catalog No. 5000238132
Encompass_Preferred

Deoxybrevianamide E is an indole alkaloid and a biosynthetic precursor for advanced metabolites isolated from marine-derived Aspergillus species. It is supplied as a research-grade small-molecule sample for natural-product and biosynthesis studies. Key identifiers include CAS 34610-68-9, molecular formula C21H25N3O2, and molecular weight 351.44 g/mol.

  • Indole alkaloid biosynthetic precursor.
  • Suitable for natural-product and biosynthesis research.
  • CAS number 34610-68-9 and molecular formula C21H25N3O2.
  • Molecular weight 351.44 g/mol.
  • Available in small quantities (for example, 1 mg) for research use.
  • Reported purity >99.0% by HPLC.
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-38132 Supplier Medchemexpress LLC Supplier No. HY1255381MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deoxybrevianamide E is an indole alkaloid and a biosynthetic precursor for advanced metabolites isolated from marine-derived Aspergillus species. It is supplied as a research-grade small-molecule sample for natural-product and biosynthesis studies. Key identifiers include CAS 34610-68-9, molecular formula C21H25N3O2, and molecular weight 351.44 g/mol.

  • Indole alkaloid biosynthetic precursor.
  • Suitable for natural-product and biosynthesis research.
  • CAS number 34610-68-9 and molecular formula C21H25N3O2.
  • Molecular weight 351.44 g/mol.
  • Available in small quantities (for example, 1 mg) for research use.
  • Reported purity >99.0% by HPLC.
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.