Learn More
Cayman Chemical DeoxycholIc ACD MaxSpec Sta
SDP
Catalog No. 502978892
Encompass_Preferred
A quantitative analytical standard guaranteed to meet MaxSpec identity, purity, stability, and concentration specifications
Catalog No. 50-297-8892 Supplier Cayman Chemical Supplier No. 31349100 UG
A quantitative analytical standard guaranteed to meet MaxSpec identity, purity, stability, and concentration specifications