Learn More
Cayman Chemical Deoxycholic Acid, Purity: ≥95%, CAS: 83-44-3, 100 g
SDP
Catalog No. 502983635
Encompass_Preferred
A secondary bile acid; inhibits TCA-induced spore germination in seven C. difficile strains at 0.2% v/v; inhibits growth and decreases the cytotoxicity of C. difficile culture supernatants to Vero cells at 0.02% v/v; inhibits ionizing radiation-induced p53-dependent transcription in a reporter assay using HCT116 cells at 200 µM; fecal and intestinal tissue levels are increased in a rat model of high-fat diet-induced obesity compared with rats fed a normal diet; increased serum levels have been found in patients with colorectal cancer
Catalog No. 50-298-3635 Supplier Cayman Chemical Supplier No. 20756100 G
A secondary bile acid; inhibits TCA-induced spore germination in seven C. difficile strains at 0.2% v/v; inhibits growth and decreases the cytotoxicity of C. difficile culture supernatants to Vero cells at 0.02% v/v; inhibits ionizing radiation-induced p53-dependent transcription in a reporter assay using HCT116 cells at 200 µM; fecal and intestinal tissue levels are increased in a rat model of high-fat diet-induced obesity compared with rats fed a normal diet; increased serum levels have been found in patients with colorectal cancer