Learn More
Research Products International Corp Deoxycholic Acid Sodium Salt, 500 G
SDP
Catalog No. 501977380
Encompass_Preferred
For solubilizing and isolating membrane-bound proteins in an active state.
Catalog No. 50-197-7380 Supplier Research Products International Corp Supplier No. D91500500.0
For solubilizing and isolating membrane-bound proteins in an active state.