Learn More
Medchemexpress LLC Deoxycholic acid | 83-44-3 | 392.57 | 1 G
SDP
Catalog No. 5000389029
Encompass_Preferred
Deoxycholic acid, also known as cholanoic acid, is a bile acid produced as a by-product of intestinal metabolism. It acts by activating the G protein-coupled bile acid receptor TGR5. This product is intended for research use only.
  • By-product of intestinal metabolism
  • Activates G protein-coupled bile acid receptor TGR5
  • Solid appearance, white to off-white
  • High purity (99.94%)
  • For research use only
Catalog No. 50-003-89029 Supplier Medchemexpress LLC Supplier No. HYN05931G
Deoxycholic acid, also known as cholanoic acid, is a bile acid produced as a by-product of intestinal metabolism. It acts by activating the G protein-coupled bile acid receptor TGR5. This product is intended for research use only.
  • By-product of intestinal metabolism
  • Activates G protein-coupled bile acid receptor TGR5
  • Solid appearance, white to off-white
  • High purity (99.94%)
  • For research use only